Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Epitope Tag Antibody (E10/V4RR) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP237825 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237825 100 μg
NB397069 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP237825 Supplier Novus Biologicals Supplier No. NBP237825
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunofluorescence.

Specifications

Antigen V5 Epitope Tag
Applications Western Blot, ELISA, Immunoprecipitation, Immunofluorescence
Classification Monoclonal
Clone E10/V4RR
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:3000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunofluorescence 1:500 - 1:2000
Formulation PBS (pH 7.2) with No Preservative
Gene Alias V5 epitope tag
Host Species Mouse
Immunogen GKPIPNPLLGLDST (V5) synthetic peptide conjugated to KLH
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epitope Tags, V5 Epitope Tags
Primary or Secondary Primary
Gene ID (Entrez) 2944
Test Specificity This product recognizes V5 (GKPIPNPLLGLDST) tagged fusion proteins. It has successfully been used in immunoprecipitation, ELISA, Western blotting, and immunofluorescence applications.
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.