Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Epitope Tag Antibody (SV5-P-K) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP25270302 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25270302 0.2 mg
NBP25270325 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25270302 Supplier Novus Biologicals Supplier No. NBP2527030.2MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

V5 Epitope Tag Monoclonal specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Radioimmunoassay, Immunofluorescence.

Specifications

Antigen V5 Epitope Tag
Applications Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Frozen), Radioimmune Assays (RIA), Immunofluorescence
Classification Monoclonal
Clone SV5-P-K
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Radioimmunoassay, Immunofluorescence 1:10 - 1:500
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias V5 epitope tag
Host Species Mouse
Immunogen Pk (V5) Epitope Tag (GKPIPNPLLGLDST).
Purification Method Protein A or G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognises a 14 amino acid sequence (GKPIPNPLLGLDST) derived from the P and V proteins of the paramyxoVirus simian virus 5.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.