Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Epitope Tag Antibody (SV5-P-K), Janelia Fluor™ 549 - Chimeric, Novus Biologicals™
SDP

Catalog No. NBP52653549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP52653549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP52653549 Supplier Novus Biologicals Supplier No. NBP252653JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence.

Specifications

Antigen V5EpitopeTag
Applications Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone SV5-P-K
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias V5 epitope tag
Host Species Rabbit
Immunogen Pk (V5) Epitope Tag (GKPIPNPLLGLDST).
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Recognises a 14 amino acid sequence (GKPIPNPLLGLDST) derived from the P and V proteins of the paramyxoVirus simian virus 5.
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Target Molecule Epitope Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.