Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Epitope Tag Antibody (SV5-Pk5) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP180562 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP180562 0.1 mg
NB018759 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP180562 Supplier Novus Biologicals Supplier No. NBP180562
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.

Specifications

Antigen V5 Epitope Tag
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SV5-Pk5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:500, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500
Gene Alias V5 epitope tag
Host Species Mouse
Immunogen A recombinant peptide corresponding to the PK tag fused to a class II - restricted T helper cell epitope.
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Epitope Tags, V5 Epitope Tags
Primary or Secondary Primary
Test Specificity Recognises a small epitope, termed Pk, present on the P/V proteins of the paramyxovirus, SV5.
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.