Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Epitope Tag Antibody (SV5-Pk5), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP180562IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP180562IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP180562IR Supplier Novus Biologicals Supplier No. NBP180562IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen V5 Epitope Tag
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone SV5-Pk5
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Gene Alias V5 epitope tag
Host Species Mouse
Immunogen A recombinant peptide corresponding to the PK tag fused to a class II - restricted T helper cell epitope.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognises a small epitope, termed Pk, present on the P/V proteins of the paramyxovirus, SV5.
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Target Molecule Epitope Tag
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.