Learn More
V5 Tag Monoclonal Antibody (4C12E11), THE™, GenScript™
Mouse Monoclonal Antibody
$229.50 - $418.20
Specifications
Antigen | V5 Tag |
---|---|
Clone | 4C12E11 |
Applications | ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-273-860
![]() |
Genscript Corporation
A01724100 |
100 μg |
Each for $229.50
|
|
|||||
50-273-861
![]() |
Genscript Corporation
A01724200 |
200 μg |
Each for $418.20
|
|
|||||
Description
Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL. The V5 epitope tag (GKPIPNPLLGLDST) is derived from a small epitope which presents on the P and V proteins of the paramyxovirus of simian virus 5 (SV5). V5 Tag Antibody is a useful tool in detection of recombinant proteins containing the V5 epitope.THE™ V5 Tag Antibody, mAb, Mouse is produced from the hybridoma resulting from fusion of SP2/0-Ag14 myeloma and B-lymphocytes obtained from mouse immunized with V5 tag peptide conjugated to KLH.
V5 Tag Monoclonal antibody specifically detects V5 Tag in Tag samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
V5 Tag | |
ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
PBS with 0.02% sodium azide; pH 7.4 | |
V5 tag peptide conjugated to KLH | |
Primary | |
-20°C or -80°C if preferred |
4C12E11 | |
Monoclonal | |
Lyophilized | |
RUO | |
simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag | |
IgG2a κ | |
Protein A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.