Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VABP Antibody (6C239.4), Biotin, Novus Biologicals™
SDP

Catalog No. NBP224876B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224876B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224876B Supplier Novus Biologicals Supplier No. NBP224876B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VABP Monoclonal specifically detects VABP in Human samples. It is validated for Western Blot.

Specifications

Antigen VABP
Applications Western Blot
Classification Monoclonal
Clone 6C239.4
Conjugate Biotin
Dilution Western Blot
Gene Accession No. Q9NZI2
Gene Alias A-type potassium channel modulatory protein 1, KChIP1, KCHIP1MGC95, Kv channel interacting protein 1, Kv channel-interacting protein 1, potassium channel interacting protein 1, Potassium channel-interacting protein 1, VABP, Vesicle APC-binding protein
Gene Symbols KCNIP1
Host Species Mouse
Immunogen Full length recombinant protein was used as the immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30820
Test Specificity This antibody is reactive to Human.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.