Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VABP Antibody, Novus Biologicals™
SDP

Catalog No. NBP219020 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219020 0.1 mg
NBP219020SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP219020 Supplier Novus Biologicals Supplier No. NBP219020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VABP Polyclonal specifically detects VABP in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot.

Specifications

Antigen VABP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. Q9NZI2
Gene Alias A-type potassium channel modulatory protein 1, KChIP1, KCHIP1MGC95, Kv channel interacting protein 1, Kv channel-interacting protein 1, potassium channel interacting protein 1, Potassium channel-interacting protein 1, VABP, Vesicle APC-binding protein
Gene Symbols KCNIP1
Host Species Rabbit
Immunogen Within the range of amino acids 1-35 (LQTKQRRPSKDKIEDELE) of human ABP protein were used as the immunogen (NP_055407).
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30820
Test Specificity The Uniprot number for VABP is Q9NZI2.
Target Species Human, Mouse, Rat, Pig, Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.