Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAC14 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB096386 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB096386 20 μL
NB096385 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB096386 Supplier Novus Biologicals Supplier No. NBP30527220UL

Rabbit Polyclonal Antibody

VAC14 Polyclonal specifically detects VAC14 in Human samples. It is validated for Western Blot.

Specifications

Antigen VAC14
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ArPIKfyve, FLJ10305, FLJ36622, MGC149816, protein VAC14 homolog, Tax1 (human T-cell leukemia virus type I) binding protein 1, Tax1 (human T-cell leukemia virus type I) binding protein 2, Tax1-binding protein 2, TAX1BP2FLJ46582, TRXMGC149815, Vac14 homolog (S. cerevisiae)
Host Species Rabbit
Immunogen Recombinant protein of human VAC14.
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55697
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.