Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAChT/SLC18A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100913480.025ML
Description
VAChT/SLC18A3 Polyclonal specifically detects VAChT/SLC18A3 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VAChT/SLC18A3 | |
Polyclonal | |
Unconjugated | |
Whole antisera with No Preservative | |
MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter | |
Rabbit | |
Unpurified | |
RUO | |
6572 | |
Reconstitute with 0.1 ml sterilized water to desired concentration. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
LYOPH | |
Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunohistochemistry-Paraffin 1:300-1:2000 | |
Q16572 | |
SLC18A3 | |
A synthetic peptide from human VACHT conjugated to an immunogenic carrier protein was used as the antigen. | |
0.025 mL | |
Primary | |
Specific for VACHT. | |
Rat, Mouse, Human | |
Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction