Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI SMA™ Remote Atrium

Catalog No. p-3765882 Shop All VAI Products
Click to view available options
Product Type:
SMA Remote Atrium Assembly
SMA Remote Atrium Bottom
SMA Remote Atrium Top
For Use With (Application):
Ideal for 18 mL filled Agar plates
Ideal for 25 mL filled Agar plates
Ideal for 32 mL filled Agar plates

Catalog No. 19-547-101

Supplier: VAI SMA316RE18

Only null left
Add to Cart
Add to Cart
Request a Quote

SMA™ Remote Atriums are designed to keep samples in places where the use of the standard SMA Atrium™ is either impractical due to size and space limitations, or the actual procedure of replacing the media plate would interfere with the manufacturing process. The SMA Remote Atrium extension tube attaches to 1/4 inch Hytrel™ tubing.

  • Easily connected to SMA Viable Monitoring Equipment
  • Constructed of 316L Stainless Steel
  • Completely sterilized by steam, heat, or ethylene oxide (ETO)
  • Used with standard media plates (90 or 100 mm) with 18, 25, or 32 mL fill levels
  • Requires a vacuum source to operate
  • Operate at an air flow of 1 CFM (28.3 LPM)

Specifications

Product Type SMA Remote Atrium Assembly
Material Stainless Steel
Connector Types 0.37 in. I.D. Connection
For Use With (Application) Ideal for 18 mL filled Agar plates
Airflow Rate 1 cfm
Overall Height (Metric) 3.81 cm
Overall Height (English) 1.5 in.
Diameter (Metric) Outer 11.43 cm
Diameter (English) Outer 4.5 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.