Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAI SMA™ Stainless Steel Atrium: Top Only
Shop All VAI Products
Click to view available options
For Use With (Application):
Ideal for 18 mL filled Agar plates
Ideal for 25 mL filled Agar plates
Ideal for 32 mL filled Agar plates
Diameter (English) Sieve:
0.25 in.
0.5 in.
Description
- Top Only, 1/2 in. diameter sieves, use with 18 mL, 25 mL, or 32 mL filled media plates
- Top Only, 1/4 in. diameter sieves, use with 18 mL, 25 mL, or 32 mL filled media plates
Specifications
Specifications
| Product Type | SMA Remote Atrium Top |
| Material | Stainless Steel |
| For Use With (Equipment) | SMA Atrium, SMA remote Atrium |
| For Use With (Application) | Ideal for 18 mL filled Agar plates |
| Overall Height (Metric) | 3.81 cm |
| Overall Height (English) | 1.5 in. |
| Diameter (Metric) Outer | 11.43 cm |
| Diameter (English) Outer | 4.5 in. |
| Diameter (Metric) Sieve | 1.27 cm |
| Diameter (English) Sieve | 0.5 in. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction