Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP-1/VAMP-2 Antibody (SP10), Novus Biologicals™
SDP

Catalog No. NBP119332 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119332 0.1 mL
NB018390 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP119332 Supplier Novus Biologicals Supplier No. NBP119332

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

VAMP-1/VAMP-2 Monoclonal specifically detects VAMP-1/VAMP-2 in Human, Mouse, Rat, Porcine, Bovine, Hamster, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen VAMP-1/VAMP-2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SP10
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:4000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS (pH 7.2), 0.1mM PMSF and 50% Glycerol with No Preservative
Gene Alias SPAX1, SYB1, VAMP1, VAMP-1, vesicle associated membrane protein 1
Gene Symbols VAMP1
Host Species Mouse
Immunogen Synaptic vesicle-containing fractions immunoprecipitated from human brain homogenates using anti-human Synaptophysin monoclonal antibodies.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6843
Test Specificity VAMP2 (SP10)
Target Species Human, Mouse, Rat, Pig, Bovine, Hamster, Xenopus species
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.