Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP-2 Rabbit anti-Human, Mouse, Rat, Clone: 8X9D9, Novus Biologicals™
SDP

Catalog No. NB167816 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167816 20 μg
NB167815 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167816 Supplier Novus Biologicals Supplier No. NBP31636220UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

VAMP-2 Monoclonal antibody specifically detects VAMP-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen VAMP-2
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 8X9D9
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias FLJ11460, SYB2vesicle-associated membrane protein 2, synaptobrevin 2, Synaptobrevin-2, VAMP-2vesicle associated membrane protein 2, vesicle-associated membrane protein 2 (synaptobrevin 2)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human VAMP-2 (P63027).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 6844
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.