Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAMP-2 Rabbit anti-Human, Mouse, Rat, Clone: 8X9D9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | VAMP-2 |
---|---|
Clone | 8X9D9 |
Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
VAMP-2 Monoclonal antibody specifically detects VAMP-2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
VAMP-2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ11460, SYB2vesicle-associated membrane protein 2, synaptobrevin 2, Synaptobrevin-2, VAMP-2vesicle associated membrane protein 2, vesicle-associated membrane protein 2 (synaptobrevin 2) | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human VAMP-2 (P63027). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
8X9D9 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Membrane Vesicle Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6844 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title