Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP-5 Antibody (3E5), FITC, Novus Biologicals™
SDP

Catalog No. NBP250357F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250357F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250357F Supplier Novus Biologicals Supplier No. NBP250357F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VAMP-5 Monoclonal specifically detects VAMP-5 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen VAMP-5
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 300000
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias myobrevin, VAMP-5, vesicle-associated membrane protein 5, vesicle-associated membrane protein 5 (myobrevin)
Gene Symbols VAMP5
Host Species Mouse
Immunogen GST-mVAMP-5-II and KLH-mVAMP-5-II peptide sequences
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Vesicle Markers, Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 10791
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.