Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP-5 Antibody (3E5), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB309544 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309544 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309544 Supplier Novus Biologicals Supplier No. NBP250357MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VAMP-5 Monoclonal antibody specifically detects VAMP-5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen VAMP-5
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3E5
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias myobrevin, VAMP-5, vesicle-associated membrane protein 5, vesicle-associated membrane protein 5 (myobrevin)
Host Species Mouse
Immunogen GST-mVAMP-5-II and KLH-mVAMP-5-II peptide sequences
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Vesicle Markers, Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 10791
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.