Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP-7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB166286 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166286 20 μg
NB166285 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166286 Supplier Novus Biologicals Supplier No. NBP31559520UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VAMP-7 Polyclonal antibody specifically detects VAMP-7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen VAMP-7
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias FLJ54296, SYBL1FLJ53762, synaptobrevin-like 1, Synaptobrevin-like protein 1, tetanus neurotoxin-insensitive VAMP, Tetanus-insensitive VAMP, Ti-VAMP, TIVAMP, VAMP-7FLJ53045, vesicle-associated membrane protein 7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 11-70 of human VAMP-7 (P51809).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Endosome Markers
Primary or Secondary Primary
Gene ID (Entrez) 6845
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.