Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP3/Cellubrevin Antibody (4G9), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N197948PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N197948PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N197948PEC5 Supplier Novus Biologicals Supplier No. NBP197948PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VAMP3/Cellubrevin Monoclonal specifically detects VAMP3/Cellubrevin in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen VAMP3/Cellubrevin
Applications Flow Cytometry
Classification Monoclonal
Clone 4G9
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CEBCellubrevin, cellubrevin, SYB3, Synaptobrevin-3, VAMP-3, vesicle-associated membrane protein 3, vesicle-associated membrane protein 3 (cellubrevin)
Gene Symbols VAMP3
Host Species Mouse
Immunogen Recombinant human cellubrevin (1-77aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 9341.0
Test Specificity Anti-human VAMP3 mAb, clone k2A2, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human cellubrevin protein.
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.