Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAMP4 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB143879 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB143879 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB143879 Supplier Novus Biologicals Supplier No. NBP305918AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VAMP4 Polyclonal specifically detects VAMP4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen VAMP4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias VAMP24, VAMP-4, vesicle-associated membrane protein 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human VAMP4 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Membrane Vesicle Markers, Neuronal Cell Markers, Neurotransmission, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 8674
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.