Learn More
Vanadium(IV) etioporphyrin III oxide
Description
Vanadium(IV) etioporphyrin III oxide is used to check the trapping efficiency of the external AlOxlayer in AlOxcoating of ultrastable Zeolite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Purple |
| Physical Form | Crystalline |
| Quantity | 50 mg |
| Solubility Information | Insoluble in water. |
| Formula Weight | 543.61 |
| Odor | Odorless |
| Chemical Name or Material | Vanadium(IV) etioporphyrin III oxide |
Safety and Handling
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.