Learn More
Vanadium(V) oxide, 99.9% (metals basis)
Description
Vanadium(V) oxide is used in Vacuum deposition, catalysts for conversion of toluene to benzonitrile or propylene to acrylonitrile, as a detector material in bolometers and microbolometer arrays for thermal imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1750°C (decomposition) |
| Melting Point | 690°C |
| Physical Form | Pieces |
| Quantity | 25 g |
| UN Number | UN2862 |
| Merck Index | 14,9921 |
| Solubility Information | Soluble in water(8g/L). |
| Formula Weight | 181.88 |
| Percent Purity | 99.9% |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H318-H335-H341-H351-H361d-H372 |
| Hazard Statement | GHS H Statement H372-H341-H361-H302-H332-H335 Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Suspected of damaging fertility or the unborn child. Harmful if swallowed. Harmful if inhaled. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM PENTOXIDE |
| EINECSNumber | 215-239-8 |
| RTECSNumber | YW2125000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.