Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Vandetanib
GREENER_CHOICE

Catalog No. p-7223004
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent VEGFR-2 inhibitor, also inhibits EGFR and RET oncoproteins activity

Vandetanib is a potent VEGFR-2 inhibitor (IC50 = 0.04 μM), and inhibitor of EGFR (IC50 = 0.5 μM). Vandetanib potently inhibits (IC50 = 100 nM) RET oncoprotein activity in human thyroid cancer. Vandetanib displays anti-angiogenic properties in human nasopharyngeal carcinoma in mice model. Vandetanib is orally available and effective in vivo.

Specifications

Chemical Name or Material N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-[(1-methyl-4-piperidinyl)methoxy]-4-quinazolinamine
CAS 443913-73-3
Quantity 10 mg
Target VEGF Receptor Inhibitors
Molecular Formula C22H24BrFN4O2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.