Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanilloid R-like 3/TRPV3 Antibody (N15/39), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP212918Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212918Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212918Z Supplier Novus Biologicals Supplier No. NBP212918AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Vanilloid R-like 3/TRPV3 Monoclonal specifically detects Vanilloid R-like 3/TRPV3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Vanilloid R-like 3/TRPV3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S15-39
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Gene Alias transient receptor potential cation channel subfamily V member 3, transient receptor potential cation channel, subfamily V, member 3, TrpV3, vanilloid receptor 3, Vanilloid receptor-like 3, vanilloid receptor-related osmotically activated channel protein, VRL3, VRL-3
Gene Symbols TRPV3
Host Species Mouse
Immunogen Synthetic peptide amino acids 458-474 (cytoplasmic C-terminus) of rat TrpV3
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162514
Test Specificity Detects approx 70kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.