Learn More
Description
Specifications
Specifications
| Antigen | Vanilloid R1/TRPV1 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:40, Immunohistochemistry (Negative) |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220 |
| Gene Symbols | TRPV1 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to an N-terminal portion of the rat TRPV1 protein (between residues 1-50) [Uniprot: O35433] |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
