Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanilloid R1/TRPV1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10098886 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10098886 0.1 mL
NB017960 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10098886 Supplier Novus Biologicals Supplier No. NB10098886
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

Vanilloid R1/TRPV1 Polyclonal specifically detects Vanilloid R1/TRPV1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Vanilloid R1/TRPV1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:1000
Formulation Lyophilized from whole antisera with No Preservative
Gene Accession No. Q8NER1
Gene Alias Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220
Gene Symbols TRPV1
Host Species Rabbit
Immunogen A synthetic peptide from the 1t cytoplasmic loop of human Vanilloid R1/TRPV1 conjugated to blue carrier protein was used as the antigen.
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Sensory Systems
Primary or Secondary Primary
Gene ID (Entrez) 7442
Reconstitution Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.