Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vanin-1/VNN1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB320809 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320809 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320809 Supplier Novus Biologicals Supplier No. NBP298364PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vanin-1/VNN1 Polyclonal antibody specifically detects Vanin-1/VNN1 in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Vanin-1/VNN1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 3.5.1, EC 3.5.1.92, HDLCQ8, MGC116930, MGC116931, MGC116932, pantetheinase, Pantetheine hydrolase, Tiff66MGC116933, vanin 1, vanin-1, vannin 1, Vascular non-inflammatory molecule 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Mouse Vanin-1/VNN1 (Uniprot#: Q9Z0K8; Met1-Ser487)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cell Biology, Endocrinology, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 8876
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.