Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAP-1/AOC3 Antibody (393106), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB303764 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303764 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303764 Supplier Novus Biologicals Supplier No. FAB39571S

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

VAP-1/AOC3 Monoclonal antibody specifically detects VAP-1/AOC3 in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen VAP-1/AOC3
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 393106
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias amine oxidase, copper containing 3 (vascular adhesion protein 1), Copper amine oxidase, EC 1.4.3, HPAOSSAO, Semicarbazide-sensitive amine oxidase, VAP1EC 1.4.3.21, VAP-1membrane primary amine oxidase, Vascular adhesion protein 1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human VAP-1/AOC3, Gly27-Asn763, Accession # Q16853
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 8639
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.