Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAP-1/AOC3 Antibody (TK8-14), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB014707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014707 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014707 Supplier Novus Biologicals Supplier No. NBP281043C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

VAP-1/AOC3 Monoclonal specifically detects VAP-1/AOC3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen VAP-1/AOC3
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone TK8-14
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias amine oxidase, copper containing 3 (vascular adhesion protein 1), Copper amine oxidase, EC 1.4.3, HPAOSSAO, Semicarbazide-sensitive amine oxidase, VAP1EC 1.4.3.21, VAP-1membrane primary amine oxidase, Vascular adhesion protein 1
Gene Symbols AOC3
Host Species Rabbit
Immunogen Affinity purified VAP-1/AOC3 from tonsil stroma of human origin.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8639
Test Specificity Recognises human Vascular Adhesion Protein-1 (VAP-1/AOC3), a glycosylated homodimeric membrane protein consisting of two 90 kDa subunits connected by disulfide bonds. Epitope recognized by TK8-18 is present both in dimeric and monomeric forms of VAP-1/AOC3. Elevated soluble VAP-1/AOC3 serum- levels have been described in several inflammatory- diseases as well as colorectal cancer. TK8-14 is a function-blocking mAb.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.