Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAP-A Antibody (OTI10E10), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP274825V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274825V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274825V Supplier Novus Biologicals Supplier No. NBP274825V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VAP-A Monoclonal specifically detects VAP-A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen VAP-A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI10E10
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias hVAP-33, MGC3745,33 kDa VAMP-associated protein, VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa, VAMP-A, VAP-33, VAP33VAMP (vesicle-associated membrane protein)-associated protein A (33kD), VAP-AVAMP-associated protein A, vesicle-associated membrane protein-associated protein A
Gene Symbols VAPA
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 2-227 of human VAP-A (NP_919415) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9218.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.