Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAP-B Antibody (736904), Biotin, Novus Biologicals™
SDP

Catalog No. NB377518 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377518 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377518 Supplier Novus Biologicals Supplier No. FAB58551B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VAP-B Monoclonal antibody specifically detects VAP-B in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF

Specifications

Antigen VAP-B
Applications Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
Classification Monoclonal
Clone 736904
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias ALS8VAMP-B/VAMP-C, VAMP (vesicle-associated membrane protein)-associated protein B and C, VAMP-associated 33 kDa protein, VAMP-associated protein B/C, VAMP-B, VAP-B, VAP-B/VAP-C, VAPC, VAP-C, vesicle-associated membrane protein-associated protein B/C
Host Species Mouse
Immunogen E. coli-derived recombinant human VAP-B, Ala2-Pro132, Accession # O95292
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9217
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.