Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cole-Parmer™ VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys
VapLock™ Solvent Waste Manifold Caps for Bottles and Carboys connect to waste containers for a secure closed system
$300.60 - $695.81
Specifications
For Use With (Application) | Solvent Safety |
---|---|
For Use With (Equipment) | Solvent-Compatible Containers |
Catalog Number | Mfr. No. | Size | Material | Thread Size (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Material | Thread Size (English) | Price | Quantity & Availability | |||
13-311-058
|
Cole-Parmer™
1201858 |
GL38 | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $370.70
|
|
|||
13-311-059
|
Cole-Parmer™
1201859 |
GL38 | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $345.95
|
|
|||
13-311-060
|
Cole-Parmer™
1201860 |
GL38 | PTFE | 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $524.29
|
|
|||
13-311-061
|
Cole-Parmer™
1201861 |
GL38 | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $450.31
|
|
|||
13-311-064
|
Cole-Parmer™
1201864 |
Nalgene 38-430 | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $377.09
|
|
|||
13-311-065
|
Cole-Parmer™
1201865 |
Nalgene 38-430 | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $302.84
|
|
|||
13-311-066
|
Cole-Parmer™
1201866 |
Nalgene 38-430 | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $484.49
|
|
|||
13-311-067
|
Cole-Parmer™
1201867 |
Nalgene 38-430 | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $564.89
|
|
|||
13-311-070
|
Cole-Parmer™
1201870 |
Wako 38 mm | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $338.18
|
|
|||
13-311-071
|
Cole-Parmer™
1201871 |
Wako 38 mm | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $323.78
|
|
|||
13-311-072
|
Cole-Parmer™
1201872 |
Wako 38 mm | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $584.66
|
|
|||
13-311-073
|
Cole-Parmer™
1201873 |
Wako 38 mm | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $497.68
|
|
|||
13-311-076
|
Cole-Parmer™
1201876 |
GL45 | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $342.70
|
|
|||
13-311-077
|
Cole-Parmer™
1201877 |
GL45 | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $350.44
|
|
|||
13-311-078
|
Cole-Parmer™
1201878 |
GL45 | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $627.02
|
|
|||
13-311-079
|
Cole-Parmer™
1201879 |
GL45 | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $632.59
|
|
|||
13-311-082
|
Cole-Parmer™
1201882 |
DIN 45 | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $347.65
|
|
|||
13-311-083
|
Cole-Parmer™
1201883 |
DIN 45 | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $301.78
|
|
|||
13-311-085
|
Cole-Parmer™
1201885 |
DIN 45 | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $621.20
|
|
|||
13-311-088
|
Cole-Parmer™
1201888 |
DIN 50 | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $300.60
|
|
|||
13-311-089
|
Cole-Parmer™
1201889 |
DIN 50 | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $304.02
|
|
|||
13-311-094
|
Cole-Parmer™
1201894 |
51 mm | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $300.67
|
|
|||
13-311-095
|
Cole-Parmer™
1201895 |
51 mm | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $336.04
|
|
|||
13-311-097
|
Cole-Parmer™
1201897 |
51 mm | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $450.38
|
|
|||
13-311-102
|
Cole-Parmer™
1201900 |
53 B | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $553.84
|
|
|||
13-311-103
|
Cole-Parmer™
1201901 |
53 B | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $417.88
|
|
|||
13-311-104
|
Cole-Parmer™
1201902 |
53 B | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $673.58
|
|
|||
13-311-105
|
Cole-Parmer™
1201903 |
53 B | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $469.75
|
|
|||
13-311-114
|
Cole-Parmer™
1201912 |
61 mm | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $388.15
|
|
|||
13-311-115
|
Cole-Parmer™
1201913 |
61 mm | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $345.96
|
|
|||
13-311-116
|
Cole-Parmer™
1201914 |
61 mm | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $484.25
|
|
|||
13-311-120
|
Cole-Parmer™
1201918 |
63 mm | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $357.02
|
|
|||
13-311-121
|
Cole-Parmer™
1201919 |
63 mm | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $333.83
|
|
|||
13-311-122
|
Cole-Parmer™
1201920 |
63 mm | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $499.91
|
|
|||
13-311-123
|
Cole-Parmer™
1201921 |
63 mm | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $584.20
|
|
|||
13-311-132
|
Cole-Parmer™
1201930 |
70 mm | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $496.39
|
|
|||
13-311-133
|
Cole-Parmer™
1201931 |
70 mm | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $437.23
|
|
|||
13-311-134
|
Cole-Parmer™
1201932 |
70 mm | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $695.81
|
|
|||
13-311-135
|
Cole-Parmer™
1201933 |
70 mm | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $469.63
|
|
|||
13-311-138
|
Cole-Parmer™
1201936 |
GL80 | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $480.81
|
|
|||
13-311-139
|
Cole-Parmer™
1201937 |
GL80 | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $402.60
|
|
|||
13-311-144
|
Cole-Parmer™
1201942 |
83 B | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $540.74
|
|
|||
13-311-145
|
Cole-Parmer™
1201943 |
83 B | Polypropylene | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $440.40
|
|
|||
13-311-146
|
Cole-Parmer™
1201944 |
83 B | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F), 1/4 in.-28 UNF(F) |
Each for $642.71
|
|
|||
13-311-147
|
Cole-Parmer™
1201945 |
83 B | PTFE | 1/2 in. NPT(F), 1/4 in. NPT(F) |
Each for $514.98
|
|
|||
Description
- Select from a variety of cap sizes to fit your container
- Different materials, numbers and sizes of ports help customize to your application
Warranty and Services
None
Specifications
Solvent Safety | |
Solvent-Compatible Containers |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title