Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Threaded Plugs

Use VapLock™ Threaded Plugs to seal unused ports for a leak-resistant system

$20.32 - $88.68

Specifications

Type Port Plug
For Use With (Application) Solvent Safety
For Use With (Equipment) VapLock Solvent Containment Systems
View More Specs

Products 8
Catalog Number Mfr. No. Material Thread Size (English) Price Quantity & Availability  
Catalog Number Mfr. No. Material Thread Size (English) Price Quantity & Availability  
13-311-249
View Documents
Cole-Parmer™
1202047
PTFE 1/4 in.-28 UNF(M)
Pack of 10 for $76.01
Only null left
Add to Cart
 
13-311-253
View Documents
Cole-Parmer™
1202051
PTFE 5/16 in.-24 UNF(M)
Each for $52.77
Only null left
Add to Cart
 
13-311-246
View Documents
Cole-Parmer™
1202044
Polypropylene 1/4 in. NPT(M)
Pack of 10 for $22.49
Only null left
Add to Cart
 
13-311-252
View Documents
Cole-Parmer™
1202050
Polypropylene 1/8 in. NPT(M)
Pack of 10 for $24.10
Only null left
Add to Cart
 
13-311-250
View Documents
Cole-Parmer™
1202048
PTFE 1/4 in. NPT(M)
Each for $88.68
Only null left
Add to Cart
 
13-311-247
View Documents
Cole-Parmer™
1202045
Polypropylene 1/2 in. NPT(M)
Pack of 10 for $29.09
Only null left
Add to Cart
 
13-311-251
View Documents
Cole-Parmer™
1202049
Polypropylene 3/8 in. NPT(M)
Pack of 10 for $20.32
Only null left
Add to Cart
 
13-311-248
View Documents
Cole-Parmer™
1202046
POM 1/4 in.-28 UNF(M)
Pack of 10 for $50.87
Only null left
Add to Cart
 
Description

Description

Seal unused manifold and bottle cap ports

Warranty and Services

None

Specifications

Specifications

Port Plug
Solvent Safety
VapLock Solvent Containment Systems
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.