Learn More
Walter Products™ Variable Volume 12 Channel Micropipettes
SCI_ED
Catalog No. S139023 Shop All Walter Products Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
10 to 100 μL
20 to 200 μL
30 to 300 μL
5 to 50 μL
Accuracy:
±0.6/1.2/3%
±0.8/1.4/4%
±0.8/1.4/6%
±1.6/2/8%
For Use With (Equipment):
20 μL Tips
200 μL Tips
200/300 μL Tips
300 μL Tips
Increments:
0.01 μL
0.05 μL
0.1 μL
0.2 μL
0.3 μL
These multichannel micropipettes are lightweight and convenient to operate, requiring little pipetting strength.
  • High chemical and UV resistance
  • Completely autoclavable at 121°C
  • 4-position volume settings and a volume display which can easily be read
  • True one-hand operation for both right and left handed operators
  • Rotating dispensing head for optimal pipetting convenience
  • Due to optimized design on the tip cone, compatible with a wide range of tips
Specifications
Accuracy ±0.8/1.4/4%
Autoclavable Autoclavable
For Use With 200/300 μL Tip
Number of Channels 12
Type Micropipette
Volume (Metric) 20 to 200 μL
For Use With (Equipment) 200/300 μL Tips
Increments 0.2 μL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.