Learn More
Eppendorf™ Varispenser™ 2x Bottle-Top Dispenser with Recirculation Valve
Shop All Eppendorf North America ProductsDescription
Ideal for dispensing aliquots of liquid from large supply bottles.
- Recirculation valve
- Standard GL 45 thread on all sizes for most common bottle threads
- Safety valve balls in discharge valve prevents leakage when discharge tube is not mounted
- Screwable air vent cap for easy assembly of drying tube or microfilter
- Oval-shaped dispenser housing for easy transportation
- PFA seal of the sliding piston prevents seizing up
- Fixation slide on interior toothed track for rapid and accurate volume adjustment
- High chemical resistance of all dispensing components
- Telescopic aspirating tubes for variable adjustment to bottle height
- Fully autoclavable without disassembly
- User adjustment option
Specifications
Specifications
| Product Type | Analog Bottletop Dispenser with Adapters |
| Channels | 1 |
| Volume (Metric) | 5.0 to 50 mL |
| Includes | Recirculation valve, adapters GL 32, GL 38, S 40, and telescopic intake tube (170 – 330 mm) |