Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vasohibin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP176579 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP176579 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP176579 Supplier Novus Biologicals Supplier No. NBP176579
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vasohibin Polyclonal specifically detects Vasohibin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen Vasohibin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_055724
Gene Alias KIAA1036vasohibin-1, VASH, vasohibin 1
Gene Symbols VASH1
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human VASH1. The immunogen is located within amino acids 290 - 340 of VASH1. Amino Acid Squence: KIGKGTGPPSPTKDRKKDV
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22846
Test Specificity VASH1 antibody is predicted to not cross-react with other vasohibin protein family members. At least two isoforms are known to exist; this antibody will recognize only the long isoform.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.