Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vasorin/SLIT-like 2 Antibody (101), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB334187 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB334187 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB334187 Supplier Novus Biologicals Supplier No. NBP290246MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Vasorin/SLIT-like 2 Monoclonal antibody specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for ELISA, ELISA

Specifications

Antigen Vasorin/SLIT-like 2
Applications ELISA
Classification Monoclonal
Clone 101
Conjugate mFluor Violet 610 SE
Dilution ELISA, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias Protein slit-like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), vasorin
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human Vasorin/SLIT-like 2 (Uniprot#: Q6EMK4, Met1-Pro575)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 114990
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.