Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vasorin/SLIT-like 2 Antibody, Novus Biologicals™
SDP

Catalog No. NB393015 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB393015 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB393015 Supplier Novus Biologicals Supplier No. NBP26857725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Vasorin/SLIT-like 2 Polyclonal antibody specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Vasorin/SLIT-like 2
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias Protein slit-like 2, SLITL2, slit-like 2, slit-like 2 (Drosophila), vasorin
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: LDVSNLSLQALPGDLSGLFPRLRLLAAARNPFNCVCPLSWFGPWVRESHVTLASPEETRCHFPPKNAGRLLLELDYADFGCPATTTTATVPTTRPVVREPTALSSSLAPT
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 114990
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.