Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VASP Antibody (IE273), Novus Biologicals™
SDP

Catalog No. NB013219 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013219 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013219 Supplier Novus Biologicals Supplier No. NBP280088
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

VASP Monoclonal specifically detects VASP in Human, Porcine, Bovine, Rabbit, Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen VASP
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone IE273
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Formulation PBS containing 2mg/ml BSA
Gene Alias vasodilator-stimulated phosphoprotein
Gene Symbols VASP
Host Species Mouse
Immunogen Purified human VASP.
Purification Method Protein A or G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 7408
Test Specificity Recognizes both the 46kDa (Ser157 dephospho-) and 50kDa (Ser157 phospho-) form of human, porcine, bovine, sheep and rabbit VASP.
Target Species Human, Pig, Bovine, Rabbit, Sheep
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.