Learn More
VAV1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305671AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The Vav gene was originally identified on the basis of its oncogenic activation during the course of gene transfer assays. The major translational product of the Vav proto-oncogene has been identified as a protein containing an array of structural motifs. Contained within its amino terminus are a helix-loop-helix domain and a leucine zipper motif similar to that of Myc family proteins; deletion of this region of p95Vav causes its oncogenic activation. In addition, p95Vav contains an SH2 domain, which could indicate its role as a substrate for tyrosine kinases. Expression of p95Vav is limited exclusively to cells of hematopoietic origin, including those of the erythroid, lymphoid and myeloid lineages.Specifications
VAV1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P15498 | |
VAV1 | |
Between 175 and 225 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
VAV1 | |
VAV | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7409 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.