Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAV1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | VAV1 |
---|---|
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-0987
![]() |
Bethyl Laboratories, Inc
A305671AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
The Vav gene was originally identified on the basis of its oncogenic activation during the course of gene transfer assays. The major translational product of the Vav proto-oncogene has been identified as a protein containing an array of structural motifs. Contained within its amino terminus are a helix-loop-helix domain and a leucine zipper motif similar to that of Myc family proteins; deletion of this region of p95Vav causes its oncogenic activation. In addition, p95Vav contains an SH2 domain, which could indicate its role as a substrate for tyrosine kinases. Expression of p95Vav is limited exclusively to cells of hematopoietic origin, including those of the erythroid, lymphoid and myeloid lineages.Specifications
VAV1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
VAV | |
VAV1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P15498 | |
7409 | |
Between 175 and 225 | |
Primary | |
4° C | |
VAV1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title