Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VCAM-1/CD106 Antibody (2691A), PE, Novus Biologicals™
SDP

Catalog No. NB295628 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295628 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295628 Supplier Novus Biologicals Supplier No. FAB8092P
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Human samples. It is validated for Flow Cytometry, Western Blot.

Specifications

Antigen VCAM-1/CD106
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2691A
Conjugate PE
Dilution Western Blot, Flow Cytometry
Formulation PBS
Gene Alias CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line CHO-derived human VCAM-1/CD106, Phe25-Glu698, Accession # P19320-1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7412
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.