Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VCAM-1/CD106 Antibody (MR106), PerCP, Novus Biologicals™
SDP

Catalog No. NBP143368P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143368P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143368P Supplier Novus Biologicals Supplier No. NBP143368PCP

Mouse Monoclonal Antibody

VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen VCAM-1/CD106
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MR106
Conjugate PerCP
Dilution Flow Cytometry
Gene Alias CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1
Gene Symbols VCAM1
Host Species Mouse
Immunogen The immunogen for this antibody was CD106.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7412
Target Species Rat
Content And Storage Store at 4C in the dark.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.