Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VCAM-1/CD106 Antibody (MVCAM A (429)), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP250620V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250620V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250620V Supplier Novus Biologicals Supplier No. NBP250620V
Add to Cart
Add to Cart

Rat Monoclonal Antibody

VCAM-1/CD106 Monoclonal specifically detects VCAM-1/CD106 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence, Immunocytochemistry.

Specifications

Antigen VCAM-1/CD106
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence, Immunocytochemistry
Classification Monoclonal
Clone MVCAM A (429)
Conjugate DyLight 405
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence, Immunocytochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1
Gene Symbols VCAM1
Host Species Rat
Immunogen Stromal cell line PA6
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7412
Test Specificity Recognizes murine vascular adhesion molecule 1 (VCAM-1), a cell surface glycoprotein that is also known as CD106. CD106 is expressed predominantly on endothelial cells and expression is up-regulated during inflammation. The ligand for CD106 is the alpha 4 subunit (CD49d) of the integrin VLA-4 (CD49d/CD29). Clone MVCAM.A(429) is reported to partially block VCAM-1 mediated functions.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.