Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VCIP135/VCPIP1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP132663 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132663 Supplier Novus Biologicals Supplier No. NBP132663
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VCIP135/VCPIP1 Polyclonal specifically detects VCIP135/VCPIP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen VCIP135/VCPIP1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias DKFZp686G038, DUBA3, EC 3.4.22.-, FLJ23132, KIAA1850FLJ60694, valosin containing protein (p97)/p47 complex interacting protein 1, valosin-containing protein (p97)/p47 complex-interacting protein p135, Valosin-containing protein p97/p47 complex-interacting protein 1, Valosin-containing protein p97/p47 complex-interacting protein p135, VCIP135deubiquitinating protein VCIP135, VCPIP1 VCIP135
Gene Symbols VCPIP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human VCPIP1. The exact sequence is proprietary.
Molecular Weight of Antigen 134 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80124
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.