Learn More
VDAC/Porin Polyclonal Antibody, Rockland™

Goat Polyclonal Antibody
$174.73 - $606.12
Specifications
Antigen | VDAC/Porin |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunofluorescence, Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1571
![]() ![]() |
Rockland Immunochemicals
600101HB2 |
100 μg |
Each for $606.12
|
|
|||||
50-199-1572
![]() ![]() |
Rockland Immunochemicals
600101HB2S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Anti-VDAC/Porin Antibody is directed against human VDAC1/Porin1 protein. A BLAST analysis was used to suggest that this antibody would react with VDAC1/Porin1 including mouse, rat, bovine, pig, and rabbit sources based on 100% homology for the immunogen sequence. Such broad reactivity makes this antibody useful as an excellent loading control.
Voltage-dependent anion channel (VDAC) proteins are abundant, pore-forming proteins belonging to the eukaryotic mitochondrial porins. It was discovered in the mitochondrial outer membrane. It also is expressed in the plasma membrane. At least three different VDAC genes have been identified in vertebrates. VDAC proteins are known to play an essential role in cellular metabolism and in the early stages of apoptosis. For example, VDAC constitutes a major pathway by which metabolites such as ADP/ATP, succinate and citrate are exchanged between the cytosol and mitochondria. And VDAC1 in the plasma membrane establishes a novel level of apoptosis regulation putatively via its redox activity.Specifications
VDAC/Porin | |
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Goat | |
Human, Mouse, Rat | |
P21796, Q60932, Q9Z2L0 | |
22333, 7416, 83529 | |
Primary | |
Antibody |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
AL033343; hVDAC1; MGC111064; Mitochondrial outer membrane protein porin 1; Mitochondrial Porin; mVDAC1; mVDAC5; N2441; OMP2; Outer mitochondrial membrane protein porin 1; Plasmalemmal porin; POR1; PORIN; Porin 31HL; Porin 31HM; porin POR1; PORIN-31-HL; rVDAC1; VD; VDAC; Vdac1; VDAC-1; Vdac5; VDAC-5; voltage dependent anion channel 1; voltage-dependent anion channel 1; Voltage-dependent anion-selective channel protein 1; Voltage-dependent anion-selective channel protein 5; YNL055C; YNL2441C | |
VDAC1 | |
-20°C, Avoid Freeze/Thaw Cycles | |
VDAC1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.