Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VDAC1 Antibody, Novus Biologicals™
SDP

Catalog No. NB100695 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100695 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100695 Supplier Novus Biologicals Supplier No. NB100695
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

VDAC1 Polyclonal specifically detects VDAC1 in Cynomolgus Monkey, Fish, Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, KnockDown, Simple Western, Western Blot.

Specifications

Antigen VDAC1
Applications ELISA, Immunohistochemistry, KnockDown, Simple Western, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Simple Western, ELISA 1:15000-1:30000, Immunohistochemistry 1:10-1:500, KnockDown Validated
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. P21796
Gene Alias hVDAC1, MGC111064, Outer mitochondrial membrane protein porin 1, Plasmalemmal porin, PORIN, Porin 31HL, Porin 31HM, PORIN-31-HL, VDAC, VDAC-1, voltage-dependent anion channel 1, voltage-dependent anion-selective channel protein 1
Gene Symbols VDAC1
Host Species Rabbit
Immunogen VDAC1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near amino acids 175-200 of Human VDAC1/Porin1. (Uniprot: P21796)
Molecular Weight of Antigen 30.77 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Mitochondrial Markers
Primary or Secondary Primary
Gene ID (Entrez) 7416
Test Specificity Anti-VDAC1 Antibody is directed against human VDAC1/Porin1 protein. A BLAST analysis was used to suggest that this antibody would react with VDAC1/Porin1 from a wide range or organisms, including avian, mammalian, aquatic and reptilian sources based on 100% homology for the immunogen sequence. Cross reactivity will occur with all forms of VDACs including VDAC1, VDAC2 (4 isoforms) and VDAC3 (2 isoforms). Such broad reactivity makes this antibody useful as an excellent loading control (mitochondrial).
Target Species Cynomolgus Monkey, Fish, Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.