Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VDAC1 Rabbit anti-Human, Mouse, Rat, Bovine, Rabbit, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB018597 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018597 0.025 mg
NBP145921 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018597 Supplier Novus Biologicals Supplier No. NBP1459210.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VDAC1 Polyclonal specifically detects VDAC1 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen VDAC1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 4 ug/mL, Immunohistochemistry 2.5 ug/mL
Formulation PBS (pH 7.2), 0.5% BSA, 30% Glycerol with 0.01% Thimerosal
Gene Accession No. P21796
Gene Alias hVDAC1, MGC111064, Outer mitochondrial membrane protein porin 1, Plasmalemmal porin, PORIN, Porin 31HL, Porin 31HM, PORIN-31-HL, VDAC, VDAC-1, voltage-dependent anion channel 1, voltage-dependent anion-selective channel protein 1
Gene Symbols VDAC1
Host Species Rabbit
Immunogen Synthetic peptide surrounding amino acid 189 of human VDAC.
Molecular Weight of Antigen 31 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Mitochondrial Markers
Primary or Secondary Primary
Gene ID (Entrez) 7416
Target Species Mouse, Rat, Human, Bovine, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.