Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VDAC1 Rabbit anti-Human, Mouse, Rat, Bovine, Rabbit, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $524.00
Specifications
| Antigen | VDAC1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5 - 4 ug/mL, Immunohistochemistry 2.5 ug/mL |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
Description
VDAC1 Polyclonal specifically detects VDAC1 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| VDAC1 | |
| Western Blot 0.5 - 4 ug/mL, Immunohistochemistry 2.5 ug/mL | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers, Mitochondrial Markers | |
| PBS (pH 7.2), 0.5% BSA, 30% Glycerol with 0.01% Thimerosal | |
| hVDAC1, MGC111064, Outer mitochondrial membrane protein porin 1, Plasmalemmal porin, PORIN, Porin 31HL, Porin 31HM, PORIN-31-HL, VDAC, VDAC-1, voltage-dependent anion channel 1, voltage-dependent anion-selective channel protein 1 | |
| VDAC1 | |
| IgG | |
| Affinity Purified | |
| 31 kDa |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| RUO | |
| Mouse, Rat, Human, Bovine, Rabbit | |
| P21796 | |
| 7416 | |
| Synthetic peptide surrounding amino acid 189 of human VDAC. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title