Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VDAC2/3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10056463SS
Description
VDAC2/3 Polyclonal specifically detects VDAC2/3 in Human, Bovine, Canine, Feline, Primate samples. It is validated for Western Blot.Specifications
VDAC2/3 | |
Polyclonal | |
Unconjugated | |
PBS containing 0.05% BSA with 0.05% Sodium Azide | |
FLJ23841, hVDAC2, Outer mitochondrial membrane protein porin 2, POR, VDAC-2, voltage-dependent anion channel 2, voltage-dependent anion-selective channel protein 2 | |
Rabbit | |
Protein G purified | |
RUO | |
7417 | |
Human, Bovine, Canine, Feline, Primate | |
Purified |
Western Blot | |
0.5 mg/mL | |
Western Blot 1-3 ug/ml | |
P45880 | |
VDAC2 | |
Amino acids 120-132 (KKSGKIKSSYKRE) of human VDAC2 protein were used as the immunogen. | |
0.025 mg | |
Primary | |
Reacts with Bovine, Cat, Chimpanzee, Dog and Human. 92% sequence identity with Mouse, Rat, Chicken and Zebrafish proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction